missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dimethylhydrazine, SPEX CertiPrep™
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
1,1-Dimethylhydrazine | 57-14-7 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
CAS | 57-14-7 |
Molecular Formula | C2H8N2 |
InChI Key | RHUYHJGZWVXEHW-UHFFFAOYSA-N |
SMILES | CN(C)N |
IUPAC Name | 1,1-dimethylhydrazine |
Molecular Weight (g/mol) | 60.1 |
PubChem CID | 5976 |
ChEBI | CHEBI:18853 |
Packaging | 2mL amber prescored glass ampule |
Solvent or Matrix | Methanol-P&T |
Show More |
Safety and Handling
Product Identifier
- 1,1-Dimethylhydrazine
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H350-May cause cancer.
- H370-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 1,1-dimethylhydrazine, methanol
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'