missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (1,1-Dimethoxyethyl)benzene, 97%

Brand:  Alfa Aesar™ H55696.06

Code : NEW

Additional Details : CAS Number : 4316-35-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15462018

  • £28.60 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

(1,1-Dimethoxyethyl)benzene
4316-35-2
166.22
MFCD00075617

Specifications

(1,1-Dimethoxyethyl)benzene
1.4900
1.008g/mL
95° to 98°C (12mmHg)
166.22
5g
4316-35-2
Liquid
97%
Colorless
MFCD00075617
Videos
SDS