missing translation for 'onlineSavingsMsg'
Learn More

1,1'-diethylferrocene 98%, ACROS Organics™

Brand:  Acros Organics 381100010

Code : 29

Additional Details : CAS Number : 7650-91-1 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10565983

  • £50.10 / 1g


Estimated Shipment
17-10-2017
Add to basket

Specifications

Chemical Identifiers

1, 1'-Diethylferrocene
97.5
≥97.5%
C14H18Fe
MFCD01862455
1273-97-8
100.0
98%
242.14

Specifications

1, 1'-Diethylferrocene
97.5
100.0
1.5770 to 1.5800 (20°C, 589nm)
Clear dark orange to dark red brown
≥97.5%
1g
242.14
1273-97-8
Authentic
Glass bottle
1.18g/mL
Liquid
98%
C14H18Fe
MFCD01862455
Documentation

Technical Documents