missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Diethyl-4,4'-carbocyanine iodide, 96%, Thermo Scientific Chemicals

Catalog Number 15421708
262.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4727-50-8
C25H25IN2
480.393
MFCD00011970
CEJANLKHJMMNQB-UHFFFAOYSA-M
cryptocyanin, cryptocyanine, kryptocyanin, kryptocyanine, cryptocyanine iodide, kryptocyanine iodide, 1,1'-diethylquino-4-carbocyanine iodide, cryptocyanine o.a. 2, 1,4'-carbocyanine iodide, 1,4'-quinocarbocyanine iodide
16219292
CHEBI:51502
(4E)-1-ethyl-4-[(Z)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;iodide
CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[I-]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4727-50-8
C25H25IN2
480.393
MFCD00011970
CEJANLKHJMMNQB-UHFFFAOYSA-M
cryptocyanin, cryptocyanine, kryptocyanin, kryptocyanine, cryptocyanine iodide, kryptocyanine iodide, 1,1'-diethylquino-4-carbocyanine iodide, cryptocyanine o.a. 2, 1,4'-carbocyanine iodide, 1,4'-quinocarbocyanine iodide
16219292
CHEBI:51502
(4E)-1-ethyl-4-[(Z)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;iodide
CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[I-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 250°C to 253°C
Color Green
Quantity 5 g
UN Number UN2811
Sensitivity Light sensitive
Formula Weight 480.38
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1,1'-Diethyl-4,4'-carbocyanine iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'