missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclobutanedicarboxylic acid, 99%, ACROS Organics™

£20.90 - £68.40

Chemical Identifiers

CAS 5445-51-2
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00001325
InChI Key CCQPAEQGAVNNIA-UHFFFAOYSA-N
Synonym 1,1-cyclobutanedicarboxylic acid, h2cbdca, 1,1-cyclobutanedicarboxylicacid, cyclobutanedicarboxylic acid, 1,1-cyclobutane dicarboxylic acid, 1,1-cyclobutanedicarboxylate, cyclobutanedicarbonic acid, zlchem 254, pubchem14151, acmc-1am73
PubChem CID 2568
ChEBI CHEBI:35691
IUPAC Name cyclobutane-1,1-dicarboxylic acid
SMILES C1CC(C1)(C(=O)O)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10704221
SDS View Documents
Acros Organics
110920050
5g Glass bottle
£20.90
5g
View alternative products
Save Up to 
»
Estimated Shipment: 04-10-2021
Please to purchase this item. Need a web account? Register with us today!
10040890
SDS View Documents
Acros Organics
110920250
25g Glass bottle
£68.40
25g
View alternative products
Save Up to 
»
Estimated Shipment: 04-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

5445-51-2
144.13
CCQPAEQGAVNNIA-UHFFFAOYSA-N
2568
cyclobutane-1,1-dicarboxylic acid
C6H8O4
MFCD00001325
1,1-cyclobutanedicarboxylic acid, h2cbdca, 1,1-cyclobutanedicarboxylicacid, cyclobutanedicarboxylic acid, 1,1-cyclobutane dicarboxylic acid, 1,1-cyclobutanedicarboxylate, cyclobutanedicarbonic acid, zlchem 254, pubchem14151, acmc-1am73
CHEBI:35691
C1CC(C1)(C(=O)O)C(=O)O

Specifications

(5 w/v% aq. soln.) clear
(5 w/v% aq. soln.) clear
0.1% max. (50°C)
0.1% max. (K.F.)
157°C to 161°C
99%
98.5
98.5% min. (GC)
C4H6(CO2H)2
09, 724
CCQPAEQGAVNNIA-UHFFFAOYSA-N
cyclobutane-1,1-dicarboxylic acid
2568
144.13
99%
1, 1-Cyclobutanedicarboxylic acid
Authentic
Glass bottle
White
5g
5445-51-2
100.0
C6H8O4
MFCD00001325
1,1-cyclobutanedicarboxylic acid, h2cbdca, 1,1-cyclobutanedicarboxylicacid, cyclobutanedicarboxylic acid, 1,1-cyclobutane dicarboxylic acid, 1,1-cyclobutanedicarboxylate, cyclobutanedicarbonic acid, zlchem 254, pubchem14151, acmc-1am73
C1CC(C1)(C(=O)O)C(=O)O
144.13
CHEBI:35691
Fine Crystalline Powder
SDS
Product Certifications