missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-Carbonyldipyrrolidine, 98%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
Description
Applications
Reactant in the ring-opening polymerization of six-and seven-membered cyclic pseudoureas.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Reactant in the ring-opening polymerization of six-and seven-membered cyclic pseudoureas.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
Color | Colorless to Yellow |
Density | 1.086 g/mL |
Boiling Point | 148°C to 150°C |
Flash Point | 113°C (235°F) |
Refractive Index | 1.513 |
Quantity | 10 g |
Beilstein | 1637732 |
Sensitivity | Light sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 168.24 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'