missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Carbonyldiimidazole, 97%, Thermo Scientific Chemicals

Catalog Number p-7022081
Click to view available options
Quantity:
10 g
50 g
250 g
530-62-1
C7H6N4O
162.15
MFCD00005286
PFKFTWBEEFSNDU-UHFFFAOYSA-N
1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole
68263
O=C(N1C=CN=C1)N1C=CN=C1
530-62-1
C7H6N4O
162.15
MFCD00005286
PFKFTWBEEFSNDU-UHFFFAOYSA-N
1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole
68263
O=C(N1C=CN=C1)N1C=CN=C1

Peptide coupling reagent1,1'-Carbonyldiimidazole acts as a coupling reagent and utilized for coupling of amino acids in order to prepare peptide in organic synthesis. It is also used in the preparation of beta-keto sulfones, sulfoxides and beta-enamino acid derivatives. It is used to convert alcohols and amines into carbamates, esters, and ureas. It is involved in the preparation of formylized imidazole by reaction with formic acid. Further, it is used in the synthesis of dipolar polyamides compounds. In addition to this, it is considered as an equivalent of phosgene and used to prepare asymmetric bis alkyl carbonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Peptide coupling reagent1,1′-Carbonyldiimidazole acts as a coupling reagent and utilized for coupling of amino acids in order to prepare peptide in organic synthesis. It is also used in the preparation of beta-keto sulfones, sulfoxides and beta-enamino acid derivatives. It is used to convert alcohols and amines into carbamates, esters, and ureas. It is involved in the preparation of formylized imidazole by reaction with formic acid. Further, it is used in the synthesis of dipolar polyamides compounds. In addition to this, it is considered as an equivalent of phosgene and used to prepare asymmetric bis alkyl carbonate.

Solubility
Soluble in dimethylformamide.

Notes
Store in a cool place. Incompatible with water, strong acids, strong oxidizing agents, strong bases and amines.
TRUSTED_SUSTAINABILITY
Melting Point 116°C to 120°C
pH 7.7
Assay Percent Range 97%
Quantity 10 g
UN Number UN3263
Beilstein 6826
Sensitivity Moisture sensitive
Merck Index 14,1819
Solubility Information Soluble in dimethylformamide.
IUPAC Name di(imidazol-1-yl)methanone
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 1,1'-Carbonyldiimidazole
Show More Show Less
compliance-icons
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'