missing translation for 'onlineSavingsMsg'
Learn More

1-(1'-BOC-Azetidin-3'-yl)pyrrolidine, Thermo Scientific™

Catalog Number 10280100
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 1019008-21-9
Packaging Amber Glass Bottle
Molecular Formula C12H22N2O2
Quantity 1 g
Synonym 1-1-boc-azetidin-3-yl pyrrolidine, tert-butyl 3-pyrrolidin-1-yl azetidine-1-carboxylate, 1-1'-boc-azetidin-3'-yl pyrrolidine, 1-1-boc-3-azetidinyl pyrrolidine, tert-butyl 3-pyrrolidinylazetidinecarboxylate, tert-butyl 3-pyrrolidin-1-yl azetidin-1-carboxylate
InChI Key YQSWCTNNHKPDNK-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CC(C1)N2CCCC2
IUPAC Name tert-butyl 3-pyrrolidin-1-ylazetidine-1-carboxylate
Molecular Weight (g/mol) 226.32
PubChem CID 42553000
Formula Weight 226.32
Percent Purity 95%
Chemical Name or Material 1-(1'-BOC-Azetidin-3'-yl)pyrrolidine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'