missing translation for 'onlineSavingsMsg'
Learn More

1,1-Bis(methylthio)-2-nitroethylene, 99%, Thermo Scientific Chemicals

Catalog Number p-3756948
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13623-94-4
C4H7NO2S2
165.24
MFCD00010443
NXGHEDHQXXXTTP-UHFFFAOYSA-N
1,1-bis methylthio-2-nitroethylene, 1,1-bis methylsulfanyl-2-nitroethene, 1,1-dimethylthio-2-nitroethylene, 1-nitro-2,2-bis methylthio ethylene, ethene, 1,1-bis methylthio-2-nitro, 1,1-bis methylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethene, 1,1-dimethylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethylene
83623
1,1-bis(methylsulfanyl)-2-nitroethene
CSC(=C[N+](=O)[O-])SC
This item is not returnable. View return policy
13623-94-4
C4H7NO2S2
165.24
MFCD00010443
NXGHEDHQXXXTTP-UHFFFAOYSA-N
1,1-bis methylthio-2-nitroethylene, 1,1-bis methylsulfanyl-2-nitroethene, 1,1-dimethylthio-2-nitroethylene, 1-nitro-2,2-bis methylthio ethylene, ethene, 1,1-bis methylthio-2-nitro, 1,1-bis methylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethene, 1,1-dimethylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethylene
83623
1,1-bis(methylsulfanyl)-2-nitroethene
CSC(=C[N+](=O)[O-])SC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 124°C to 126°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NCH=C(SCH3)2
Quantity 5 g
Formula Weight 165.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1, 1-Bis(methylthio)-2-nitroethylene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'