missing translation for 'onlineSavingsMsg'
Learn More

1,1-Bis(hydroxymethyl)cyclopropane, 90%, ACROS Organics™

£28.78 - £80.42

Chemical Identifiers

CAS 39590-81-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00464207
InChI Key YAINYZJQSQEGND-UHFFFAOYSA-N
Synonym 1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f
PubChem CID 4101437
IUPAC Name [1-(hydroxymethyl)cyclopropyl]methanol
SMILES C1CC1(CO)CO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10547454
SDS View Documents
Acros Organics
426250010
1g Glass bottle £28.78
1g View alternative products
Save Up to 
»
Estimated Shipment
02-11-2020
Please to purchase this item. Need a web account? Register with us today!
10031084
SDS View Documents
Acros Organics
426250050
5g Glass bottle £80.42
5g View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

39590-81-3
102.13
YAINYZJQSQEGND-UHFFFAOYSA-N
4101437
C1CC1(CO)CO
C5H10O2
MFCD00464207
1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f
[1-(hydroxymethyl)cyclopropyl]methanol

Specifications

1, 1-Bis(hydroxymethyl)cyclopropane
110°C
1.065
1g
88.0
90%
MFCD00464207
YAINYZJQSQEGND-UHFFFAOYSA-N
[1-(hydroxymethyl)cyclopropyl]methanol
4101437
90%
1.0650g/mL
Glass bottle
215.0°C to 233.0°C
39590-81-3
100.0
C5H10O2
1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f
C1CC1(CO)CO
102.13
102.13
Safety and Handling

Safety and Handling

Product Identifier
  • 1,1-Bis(hydroxymethyl)cyclopropane

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.