missing translation for 'onlineSavingsMsg'
Learn More

[1,1'-Bis(diphenylphosphino)ferrocene]tetracarbonyltungsten, 98%, Thermo Scientific™

Catalog Number p-7059266
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
67292-26-6
C38H28FeO4P2W
850.27
MFCD03427160
[1,1'-Bis(diphenylphosphino)ferrocene]tetracarbonyltungsten
[Fe].[W].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
67292-26-6
C38H28FeO4P2W
850.27
MFCD03427160
[1,1'-Bis(diphenylphosphino)ferrocene]tetracarbonyltungsten
[Fe].[W].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 220°C (decomposition)
Quantity 1 g
UN Number UN3466
Sensitivity Air sensitive
Formula Weight 850.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material [1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonyltungsten
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'