missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ [1,1'-Bis(diphenylphosphino)ferrocene]tetracarbonylmolybdenum, 98%

Brand:  Alfa Aesar™ 044281.03

Code : NEW

Additional Details : CAS Number : 67292-28-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11349818

  • £20.60 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

[1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonylmolybdenum
C38H28FeMoO4P2
MFCD03427159
67292-28-8
762.37

Specifications

[1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonylmolybdenum
Powder
C38H28FeMoO4P2
MFCD03427159
1g
67292-28-8
98%
762.37
256°C (dec.)
Videos
SDS