missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ [1,1'-Bis(diphenylphosphino)ferrocene]tetracarbonylchromium, 99%

Brand:  Alfa Aesar™ 044289.03

Code : NEW

Additional Details : CAS Number : 67292-31-3 Weight : 0.00100kg Transport : UN number : 3466 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11399828

  • £35.19 / 1g


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

[1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonylchromium
C38H28CrFeO4P2
MFCD03427158
67292-31-3
725.37
UN3466

Specifications

[1,1'-Bis (diphenylphosphino)ferrocene]tetracarbonylchromium
Powder
C38H28CrFeO4P2
MFCD03427158
136° to 139°C (dec.)
Air Sensitive
67292-31-3
99%
725.37
UN3466
1g
Videos
SDS