missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, Thermo Scientific Chemicals

Catalog Number p-3735631
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
95464-05-4, 67-56-1
C35H30Cl4FeP2Pd
816.64
MFCD00792899
Bis(diphenylphosphino)ferrocene]palladium(II) chloride
[Fe].ClCCl.Cl[Pd]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
95464-05-4, 67-56-1
C35H30Cl4FeP2Pd
816.64
MFCD00792899
Bis(diphenylphosphino)ferrocene]palladium(II) chloride
[Fe].ClCCl.Cl[Pd]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown-Red to Orange
Elemental Analysis 50 to 53% (C)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Synonym ethylidene chloride, ethane, 1,1-dichloro, ethylidene dichloride, dichloroethane, 1,1-dichlorethane, aethylidenchlorid, 1,1-dicloroetano, 1,1-ethylidene dichloride, dichloromethylmethane, rcra waste number u076
InChI Key SCYULBFZEHDVBN-UHFFFAOYSA-N
PubChem CID 6365
ChEBI CHEBI:81599
Formula Weight 816.22
Physical Form Crystalline Powder
Chemical Name or Material 1, 1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'