missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(diphenylphosphino)ferrocene, 98%, ACROS Organics™

Brand:  Acros Organics 348010050

Code : 29

Additional Details : CAS Number : 12150-46-8 Weight : 0.06000kg Transport : UN number : 3467 Chem class : 1 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10189931

  • £49.80 / 5g


Estimated Shipment
23-02-2018
Add to basket


Specifications

Chemical Identifiers

1, 1'-Bis(diphenylphosphino)ferrocene
97.5
≥97.5%
C34H28FeP2
MFCD00001422
12150-46-8
100.0
98%
553.99

Specifications

1, 1'-Bis(diphenylphosphino)ferrocene
97.5
Authentic
Powder
98%
553.99
15, 3490
5g
12150-46-8
100.0
Glass bottle
≥97.5%
C34H28FeP2
MFCD00001422
Orange
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,1``-Bis(diphenylphosphino)ferrocene

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

SDS