missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,1-Bis(diphenylphosphino)ethylene, 98%

Brand:  Alfa Aesar™ H55864.03

Code : NEW

Additional Details : CAS Number : 84494-89-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15422248

  • £57.56 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

1,1-Bis(diphenylphosphino)ethylene
[(C6H5)2P]2C=CH2
MFCD00191809
84494-89-3
396.41

Specifications

1,1-Bis(diphenylphosphino)ethylene
Solid
[(C6H5)2P]2C=CH2
MFCD00191809
1g
84494-89-3
98%
396.41
110° to 111°C
Videos
SDS