1,1'-Bis(diisopropylphosphino)ferrocene, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 363870025

UNSPSC: 12352100

Code: 29

Additional Details:
Additional Details: CAS Number: 97239-80-0 Weight: 0.00250kg


Packaging Glass bottle
Quantity 2.5g

Product Code. 10085193

Quantity Price
1 £ 158.5 / 2.50g
Estimated Shipment
25-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1, 1'-Bis(diisopropylphosphino)ferrocene
Name Note 98%
CAS 97239-80-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C22H36FeP2
Formula Weight 418.32
MDL Number MFCD02684559

Specification

Formula Weight 418.32
Physical Form Crystalline Powder
Assay ≥97.5%
Quantity 2.5g
Color Orange
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 97239-80-0
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD02684559
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Packaging Glass bottle
Molecular Formula C22H36FeP2
Name Note 98%
CAS Max % 100.0
TSCA TSCA
Chemical Name or Material 1, 1'-Bis(diisopropylphosphino)ferrocene
Melting Point 53° to 57°C
Documentation

Technical documents

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.