1,1'-Bis(di-tert-butylphosphino)ferrocene, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 429710010

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 84680-95-5 Weight: 0.05100kg


Packaging Glass bottle
Quantity 1g

Product Code. 10247857

Quantity Price
1 £ 54.3 / 1g
EU Stock 3
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1, 1'-Bis(Di-tert-butylphosphino)ferrocene
CAS 84680-95-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C26H44FeP2
Formula Weight 474.44
MDL Number MFCD01630818

Specification

Physical Form Powder
Formula Weight 474.44
Quantity 1g
Color Dark orange to red
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
NMR consistent with structure
CAS 84680-95-5
Infrared Spectrum Authentic
MDL Number MFCD01630818
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Elemental Analysis 64.18 to 67.47% C
Packaging Glass bottle
Molecular Formula C26H44FeP2
CAS Max % 100.0
Chemical Name or Material 1, 1'-Bis(Di-tert-butylphosphino)ferrocene
Melting Point 73° to 76°C
Documentation

Technical documents

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.