missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(di-tert-butylphosphino)ferrocene, 98%, ACROS Organics™

Brand:  Acros Organics 429712500

Code : 29

Additional Details : CAS Number : 84680-95-5 Weight : 0.05025kg


Quantity 250mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10165867

  • £22.60 / 250mg


Estimated Shipment
09-02-2018
Add to basket


Specifications

Chemical Identifiers

1,1′-Bis(di-tert-butylphosphino)ferrocene
97.5
98%
474.44
84680-95-5
100.0
C26H44FeP2
MFCD01630818

Specifications

1,1′-Bis(di-tert-butylphosphino)ferrocene
97.5
64.18 to 67.47% C
consistent with structure
Powder
98%
474.44
Dark orange to red
250mg
84680-95-5
100.0
Authentic
Glass bottle
98%
C26H44FeP2
MFCD01630818
73.0° to 75.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,1'-Bis(di-tert-butylphosphino)ferrocene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS