missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-1,1'-Bi(2-naphthol), 99%, Thermo Scientific Chemicals
18.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
(±)-1,1'-Bi(2-naphthol) acts as chiral separation agents. It is widely used in chiral liquid crystal,medicines,spices etc,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+/-)-1,1′-Bi(2-naphthol) acts as chiral separation agents. It is widely used in chiral liquid crystal,medicines,spices etc,.
Solubility
Soluble in dioxane (50 mg/mL). Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Refrigerated (2-8°C)
(+/-)-1,1′-Bi(2-naphthol) acts as chiral separation agents. It is widely used in chiral liquid crystal,medicines,spices etc,.
Solubility
Soluble in dioxane (50 mg/mL). Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Refrigerated (2-8°C)
Specifications
Melting Point | 215°C to 218°C |
Density | 1.301 |
Odor | Characteristic |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 997518 |
Merck Index | 14,1226 |
Solubility Information | Soluble in dioxane (50mg/mL). Insoluble in water. |
Formula Weight | 286.33 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'