missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Benzothiophen-2-yl)-2-bromo-1-ethanone, ≥97%, Thermo Scientific™

Catalog Number p-3109997
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
97511-06-3
C10H7BrOS
255.13
MFCD06658971
WVTSOGFICBVCRE-UHFFFAOYSA-N
1-1-benzothiophen-2-yl-2-bromo-1-ethanone, 1-1-benzothiophen-2-yl-2-bromoethanone, 2-bromoacetyl benzo b thiophene, 1-1-benzothiophen-2-yl-2-bromoethan-1-one, 1-benzo b thiophen-2-yl-2-bromoethanone, 1-benzo b thiophen-2-yl-2-bromoethan-1-one, 1-benzothiophen-2-yl-2-bromoethan-1-one, 2-bromoacetyl thianaphthene
2795258
1-(1-benzothiophen-2-yl)-2-bromoethanone
BrCC(=O)C1=CC2=CC=CC=C2S1
This item is not returnable. View return policy
97511-06-3
C10H7BrOS
255.13
MFCD06658971
WVTSOGFICBVCRE-UHFFFAOYSA-N
1-1-benzothiophen-2-yl-2-bromo-1-ethanone, 1-1-benzothiophen-2-yl-2-bromoethanone, 2-bromoacetyl benzo b thiophene, 1-1-benzothiophen-2-yl-2-bromoethan-1-one, 1-benzo b thiophen-2-yl-2-bromoethanone, 1-benzo b thiophen-2-yl-2-bromoethan-1-one, 1-benzothiophen-2-yl-2-bromoethan-1-one, 2-bromoacetyl thianaphthene
2795258
1-(1-benzothiophen-2-yl)-2-bromoethanone
BrCC(=O)C1=CC2=CC=CC=C2S1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 255.13
Percent Purity 97%
Chemical Name or Material 1-(1-Benzothiophen-2-yl)-2-bromo-1-ethanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'