missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(1-Adamantyl)ethylamine hydrochloride, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 215.77 |
Percent Purity | 95% |
Chemical Name or Material | 1-(1-Adamantyl)ethylamine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'