missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,4-Tetraphenyl-1,3-butadiene, 99%, ACROS Organics™

Brand:  Acros Organics 176540100

Code : 29

Additional Details : CAS Number : 1450-63-1 Weight : 0.06000kg

Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10606334

  • £97.20 / 10g


Estimated Shipment
15-06-2018
Add to basket

Specifications

Chemical Identifiers

1,1,4,4-Tetraphenyl-1,3-butadiene
1450-63-1
100.0
99%
(C6H5)2C=CHCH=C(C6H5)2
MFCD00004766
99%
98.5
99%
C28H22
358.47
TPB

Specifications

1,1,4,4-Tetraphenyl-1,3-butadiene
196.0° to 200.0°C
99%
98.5
Authentic
Needle-Like Crystals
99%
(C6H5)2C=CHCH=C(C6H5)2
MFCD00004766
TPB
White to slightly yellow
10g
1450-63-1
100.0
Glass bottle
99%
C28H22
358.47
05, 750
Videos
SDS