1-((1,3-Dioxolan-2-yl)methyl)-1H-pyrazole-4-boronic acid pinacol ester, 97%, Acros Organics


Overview
Brand: Acros Organics

Manufacturer Part Number: 444990050

UNSPSC: 12161500

Code: 29

Additional Details:
Additional Details: CAS Number: 864754-17-6 Weight: 0.05500kg


Quantity 5g

Product Code. 11906241

Quantity Price
1 £ 917.3 / 5g
Estimated Shipment date
from Supplier 12-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-((1, 3-Dioxolan-2-yl)methyl)-1H-pyrazole-4-boronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C13H21BN2O4
Formula Weight 280.13

Specification

Formula Weight 280.13
Quantity 5g
CAS 864754-17-6
Assay Percent Range 97%
Molecular Formula C13H21BN2O4
Chemical Name or Material 1-((1, 3-Dioxolan-2-yl)methyl)-1H-pyrazole-4-boronic acid pinacol ester
Documentation

Technical documents