missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(1,3-Benzothiazol-2-yl)-2-bromo-1-ethanone, 90+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Assay Percent Range | 90% |
Quantity | 1 g |
IUPAC Name | 1-(1,3-benzothiazol-2-yl)-2-bromoethanone |
Formula Weight | 256.12 |
Percent Purity | 90% |
Chemical Name or Material | 1-(1, 3-Benzothiazol-2-yl)-2-bromo-1-ethanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'