missing translation for 'onlineSavingsMsg'
Learn More

1-(1,3-Benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione, 97%, Thermo Scientific™

Catalog Number p-3484551
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
306935-39-7
C11H7F3O4
260.168
MFCD01570563
FDLJUCOQPAGQNY-UHFFFAOYSA-N
1-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1-2h-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 3-3,4-methylenedioxy benzoyl-1,1,1-trifluoroacetone, 1-benzo d 1,3 dioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, maybridge1_005379, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dion, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1,3-butanedione,1-1,3-benzodioxol-5-yl-4,4,4-trifluoro, 1-1,3-benzodioxol-5-yl-4,4,4-tri-fluorobutane-1,3-dione, 1-1,3-benzodioxole-5-yl-4,4,4-trifluorobutane-1,3-dione
2775368
1-(1,3-benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione
C1OC2=C(O1)C=C(C=C2)C(=O)CC(=O)C(F)(F)F
This item is not returnable. View return policy
306935-39-7
C11H7F3O4
260.168
MFCD01570563
FDLJUCOQPAGQNY-UHFFFAOYSA-N
1-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1-2h-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 3-3,4-methylenedioxy benzoyl-1,1,1-trifluoroacetone, 1-benzo d 1,3 dioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, maybridge1_005379, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dion, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1,3-butanedione,1-1,3-benzodioxol-5-yl-4,4,4-trifluoro, 1-1,3-benzodioxol-5-yl-4,4,4-tri-fluorobutane-1,3-dione, 1-1,3-benzodioxole-5-yl-4,4,4-trifluorobutane-1,3-dione
2775368
1-(1,3-benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione
C1OC2=C(O1)C=C(C=C2)C(=O)CC(=O)C(F)(F)F
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 260.17
Percent Purity 97%
Chemical Name or Material 1-(1,3-Benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'