missing translation for 'onlineSavingsMsg'
Learn More

(±)-1-(1,2-Diphenylethyl)piperidine maleate, Tocris Bioscience™

NMDA antagonist; acts at ion channel site

Brand:  Tocris (Bio-Techne) 036010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13457583

  • £99.96 / 10mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

High affinity antagonist at the ion channel on the NMDA receptor.
Specifications

Chemical Identifiers

207461-99-2
381.472
+/--1-1,2-diphenylethyl piperidine maleate, a+/--1-1,2-diphenylethyl piperidine maleate, piperidine, 1-1,2-diphenylethyl-, 2z-2-butenedioate, but-2-enedioic acid-1-1,2-diphenylethyl piperidine 1/1
but-2-enedioic acid;1-(1,2-diphenylethyl)piperidine
C23H27NO4
AEHZEDLXDLPQQD-UHFFFAOYSA-N
57369670
C1CCN(CC1)C(CC2=CC=CC=C2)C3=CC=CC=C3.C(=CC(=O)O)C(=O)O

Specifications

10mg
207461-99-2
+/--1-1,2-diphenylethyl piperidine maleate, a+/--1-1,2-diphenylethyl piperidine maleate, piperidine, 1-1,2-diphenylethyl-, 2z-2-butenedioate, but-2-enedioic acid-1-1,2-diphenylethyl piperidine 1/1
C1CCN(CC1)C(CC2=CC=CC=C2)C3=CC=CC=C3.C(=CC(=O)O)C(=O)O
381.472
381.47
(±)-1-(1, 2-Diphenylethyl)piperidine maleate
C23H27NO4
AEHZEDLXDLPQQD-UHFFFAOYSA-N
but-2-enedioic acid;1-(1,2-diphenylethyl)piperidine
57369670
>98%
Videos
Documents