missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane, 98.5%, Thermo Scientific Chemicals

Catalog Number p-196239
14.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


12.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
79-34-5
MFCD00000848
QPFMBZIOSGYJDE-UHFFFAOYSA-N
s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan
6591
CHEBI:36026
1,1,2,2-tetrachloroethane
C(C(Cl)Cl)(Cl)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
79-34-5
MFCD00000848
QPFMBZIOSGYJDE-UHFFFAOYSA-N
s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan
6591
CHEBI:36026
1,1,2,2-tetrachloroethane
C(C(Cl)Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point -43.0°C
Color Colorless
Density 1.5800g/mL
Boiling Point 147.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5%
Packaging Glass bottle
Molecular Formula C2H2Cl4
Linear Formula Cl2CHCHCl2
Refractive Index 1.4930 to 1.4950
Quantity 25 mL
Beilstein 01, 86
Merck Index 15, 9334
Specific Gravity 1.58
Solubility Information Solubility in water: 0.3 g/100mL (25°C). Other solubilities: miscible with methanol, ethanol, benzene, ether,, petroleum ether, carbon tetrachloride, chloroform,, carbon disulfide, dimethylformamide and oils, soluble in acetone
Molecular Weight (g/mol) 167.838
Formula Weight 167.85
Percent Purity 98.5%
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrachloroethane, 98.5%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,2,2-Tetrachloroethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H310-Fatal in contact with skin.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'