missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrabromoethane, 98%, Thermo Scientific Chemicals

Catalog Number p-3830540
23.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
79-27-6
C2H2Br4
345.65
MFCD00000133
QXSZNDIIPUOQMB-UHFFFAOYSA-N
acetylene tetrabromide, muthmann's liquid, ethane, 1,1,2,2-tetrabromo, tetrabromoacetylene, sym-tetrabromoethane, s-tetrabromoethane, 1,1,2,2-tetrabromaethan, 1,1,2,2-tetrabromoetano, tbe, unii-v00gkc584o
6588
1,1,2,2-tetrabromoethane
BrC(Br)C(Br)Br
This item is not returnable. View return policy
79-27-6
C2H2Br4
345.65
MFCD00000133
QXSZNDIIPUOQMB-UHFFFAOYSA-N
acetylene tetrabromide, muthmann's liquid, ethane, 1,1,2,2-tetrabromo, tetrabromoacetylene, sym-tetrabromoethane, s-tetrabromoethane, 1,1,2,2-tetrabromaethan, 1,1,2,2-tetrabromoetano, tbe, unii-v00gkc584o
6588
1,1,2,2-tetrabromoethane
BrC(Br)C(Br)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 1°C
Density 2.9600g/mL
Boiling Point 244°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Br2CHCHBr2
Refractive Index 1.6355 to 1.638
Quantity 250 g
Beilstein 01, 94
Merck Index 15, 9330
Specific Gravity 2.96
Solubility Information Solubility in water: 0.63g/L (20°C). Other solubilities: soluble in acetone,benzene,miscible with ethanol,ether,chloroform,aniline,and glacial acetic acid
Formula Weight 345.64
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrabromoethane
Show More Show Less
compliance-icons
Product Identifier
  • 1,1,2,2-Tetrabromoethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,1,2,2-Tetrabromoethane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'