missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Tris(hydroxymethyl)ethane, 97%, Thermo Scientific Chemicals

Catalog Number p-209075
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
77-85-0
C5H12O3
120.15
MFCD00004687
QXJQHYBHAIHNGG-UHFFFAOYSA-N
trimethylolethane, 1,1,1-tris hydroxymethyl ethane, trimet, methriol, metriol, pentaglycerine, pentaglycerol, methyltrimethanolmethane, 2-hydroxymethyl-2-methylpropane-1,3-diol, tris hydroxymethyl ethane
6502
2-(hydroxymethyl)-2-methylpropane-1,3-diol
CC(CO)(CO)CO
This item is not returnable. View return policy
77-85-0
C5H12O3
120.15
MFCD00004687
QXJQHYBHAIHNGG-UHFFFAOYSA-N
trimethylolethane, 1,1,1-tris hydroxymethyl ethane, trimet, methriol, metriol, pentaglycerine, pentaglycerol, methyltrimethanolmethane, 2-hydroxymethyl-2-methylpropane-1,3-diol, tris hydroxymethyl ethane
6502
2-(hydroxymethyl)-2-methylpropane-1,3-diol
CC(CO)(CO)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 200.0°C to 203.0°C
Color White
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C(CH2OH)3
Quantity 25 g
Beilstein 01, 520
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols, insoluble in ether, benzene
Formula Weight 120.15
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1, 1, 1-Tris(hydroxymethyl)ethane, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1,1,1-Tris(hydroxymethyl)ethane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'