missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-2-propanol, 97%, Thermo Scientific Chemicals

Catalog Number 11367586
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
374-01-6
C3H5F3O
114.067
MFCD00014403
GILIYJDBJZWGBG-UHFFFAOYSA-N
1,1,1-trifluoro-2-propanol, 2-propanol, 1,1,1-trifluoro, 1,1,1-trifluoroisopropanol, 1,1,1-trifluoropropanol-2, 1,1,1-trifluoro-propan-2-ol, 1-methyl-2,2,2-trifluoroethanol, pubchem12648, acmc-20ap8u, 1,1-trifluoro-2-propanol, 2-propanol,1,1-trifluoro
9774
1,1,1-trifluoropropan-2-ol
CC(C(F)(F)F)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
374-01-6
C3H5F3O
114.067
MFCD00014403
GILIYJDBJZWGBG-UHFFFAOYSA-N
1,1,1-trifluoro-2-propanol, 2-propanol, 1,1,1-trifluoro, 1,1,1-trifluoroisopropanol, 1,1,1-trifluoropropanol-2, 1,1,1-trifluoro-propan-2-ol, 1-methyl-2,2,2-trifluoroethanol, pubchem12648, acmc-20ap8u, 1,1-trifluoro-2-propanol, 2-propanol,1,1-trifluoro
9774
1,1,1-trifluoropropan-2-ol
CC(C(F)(F)F)O

1,1,1-Trifluoro-2-propanol is used as a chemical building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trifluoro-2-propanol is used as a chemical building block.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.259
Boiling Point 75°C to 76°C
Flash Point 25°C (77°F)
Refractive Index 1.3175
Quantity 25 g
UN Number UN1987
Beilstein 1734738
Solubility Information Fully miscible in water.
Formula Weight 114.07
Percent Purity 97%
Chemical Name or Material 1,1,1-Trifluoro-2-propanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'