missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1-Trifluoro-2-propanol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1,1,1-Trifluoro-2-propanol is used as a chemical building block.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1,1-Trifluoro-2-propanol is used as a chemical building block.
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
1,1,1-Trifluoro-2-propanol is used as a chemical building block.
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications
Density | 1.259 |
Boiling Point | 75°C to 76°C |
Flash Point | 25°C (77°F) |
Refractive Index | 1.3175 |
Quantity | 5 g |
UN Number | UN1987 |
Beilstein | 1734738 |
Solubility Information | Fully miscible in water. |
Formula Weight | 114.07 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'