missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate converts benzisoxazole to α-aryloxyisobutyric acid. It forms eutectic with dimethyl sulfone, which is the most suitable media for freeze-drying due to its high solubilizing ability and a good rate of solvent removal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate converts benzisoxazole to α-aryloxyisobutyric acid. It forms eutectic with dimethyl sulfone, which is the most suitable media for freeze-drying due to its high solubilizing ability and a good rate of solvent removal.
Solubility
Soluble in ethanol, ether, chloroform, and glycerol. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate converts benzisoxazole to α-aryloxyisobutyric acid. It forms eutectic with dimethyl sulfone, which is the most suitable media for freeze-drying due to its high solubilizing ability and a good rate of solvent removal.
Solubility
Soluble in ethanol, ether, chloroform, and glycerol. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
Melting Point | 98°C to 100°C (anhydrous) |
Boiling Point | 167°C |
Flash Point | >100°C (212°F) |
Quantity | 25 g |
Beilstein | 878167 |
Merck Index | 14,2129 |
Solubility Information | Soluble in ethanol,ether,chloroform,and glycerol. Insoluble in water. |
Formula Weight | 186.47 (177.46 Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | 1,1,1-Trichloro-2-methyl-2-propanol hemihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'