missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, 98+%
£30.24 - £383.00
Chemical Identifiers
CAS | 1522-22-1 |
---|---|
Molecular Formula | C5H2F6O2 |
Molecular Weight (g/mol) | 208.059 |
MDL Number | MFCD00000426 |
InChI Key | QAMFBRUWYYMMGJ-UHFFFAOYSA-N |
Synonym | hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hfac, 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,3-bis trifluoromethyl propane-1,3-dione, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane |
PubChem CID | 73706 |
IUPAC Name | 1,1,1,5,5,5-hexafluoropentane-2,4-dione |
SMILES | C(C(=O)C(F)(F)F)C(=O)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351377
|
Alfa Aesar™
L01693.06 |
5g |
£30.24
5g |
In stock, ready to ship: 33 | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11361377
|
Alfa Aesar™
L01693.14 |
25g |
£124.00
25g |
In stock, ready to ship: 28 | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11371377
|
Alfa Aesar™
L01693.22 |
100g |
£383.00
100g |
In stock, ready to ship: 18 | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Applications1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.
Solubility
Not miscible in water.
Notes
Store away from oxidizing agents, reducing agents and water. Hygroscopic.
Chemical Identifiers
1522-22-1 | |
208.059 | |
QAMFBRUWYYMMGJ-UHFFFAOYSA-N | |
73706 | |
C(C(=O)C(F)(F)F)C(=O)C(F)(F)F |
C5H2F6O2 | |
MFCD00000426 | |
hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hfac, 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,3-bis trifluoromethyl propane-1,3-dione, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane | |
1,1,1,5,5,5-hexafluoropentane-2,4-dione |
Specifications
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione | |
32°C (89°F) | |
Hygroscopic | |
5g | |
C5H2F6O2 | |
UN2920 | |
hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hfac, 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,3-bis trifluoromethyl propane-1,3-dione, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane | |
QAMFBRUWYYMMGJ-UHFFFAOYSA-N | |
1,1,1,5,5,5-hexafluoropentane-2,4-dione | |
73706 | |
≥98% |
1.486 | |
1.334 | |
69°C to 71°C | |
1522-22-1 | |
MFCD00000426 | |
1790138 | |
Not miscible in water. | |
C(C(=O)C(F)(F)F)C(=O)C(F)(F)F | |
208.059 | |
208.06 |