missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.
Solubility
Not miscible in water.
Notes
Store away from oxidizing agents, reducing agents and water. Hygroscopic.
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.
Solubility
Not miscible in water.
Notes
Store away from oxidizing agents, reducing agents and water. Hygroscopic.
Specifications
Density | 1.486 |
Boiling Point | 69°C to 71°C |
Flash Point | 32°C (89°F) |
Refractive Index | 1.334 |
Quantity | 5 g |
UN Number | UN2920 |
Beilstein | 1790138 |
Sensitivity | Hygroscopic |
Solubility Information | Not miscible in water. |
Formula Weight | 208.06 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'