missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexamethyldisilazane, 98%, Thermo Scientific Chemicals

Catalog Number p-4687108
Click to view available options
Quantity:
10 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
999-97-3
C6H19NSi2
161.4
FFUAGWLWBBFQJT-UHFFFAOYSA-N
hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
13838
CHEBI:85068
[dimethyl-(trimethylsilylamino)silyl]methane
C[Si](C)(C)N[Si](C)(C)C
This item is not returnable. View return policy
999-97-3
C6H19NSi2
161.4
FFUAGWLWBBFQJT-UHFFFAOYSA-N
hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
13838
CHEBI:85068
[dimethyl-(trimethylsilylamino)silyl]methane
C[Si](C)(C)N[Si](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -78.0°C
Color Colorless
Density 0.7600g/mL
Boiling Point 125.0°C
Flash Point 20°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiNHSi(CH3)3
Refractive Index 1.4060 to 1.4090
Quantity 10 mL
Merck Index 15,4725
Specific Gravity 0.76
Solubility Information Solubility in water: reacts. Other solubilities: reacts with methanol, soluble in acetone, benzene, ethyl ether, pentane, and perchloroethylene
Viscosity 0.9 mm2/s (25°C)
Formula Weight 161.4
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexamethyldisilazane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1,1,3,3,3-Hexamethyldisilazane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'