missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,1,1,3,3,3-Hexafluoroisopropyl p-toluenesulfonate, 97%

Brand:  Alfa Aesar™ L10864.06

Code : NEW

Additional Details : CAS Number : 67674-48-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11343046

  • £19.30 / 5g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

1,1,1,3,3,3-Hexafluoroisopropyl p-toluenesulfonate
C10H8F6O3S
MFCD00039262
p-Toluenesulfonic acid 1,1,1,3,3,3-hexafluoroisopropyl ester
67674-48-0
322.23
4530866

Specifications

1,1,1,3,3,3-Hexafluoroisopropyl p-toluenesulfonate
5g
97%
322.23
4530866
39° to 40°C
67674-48-0
C10H8F6O3S
MFCD00039262
p-Toluenesulfonic acid 1,1,1,3,3,3-hexafluoroisopropyl ester
Videos
SDS