missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropanol-d2, 99+ atom% D, Thermo Scientific Chemicals

Catalog Number 10582071
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
38701-74-5
C3H2F6O
170.05
MFCD00037559
BYEAHWXPCBROCE-AWPANEGFSA-N
2724852
2-deuterio-2-deuteriooxy-1,1,1,3,3,3-hexafluoropropane
[2H]OC([2H])(C(F)(F)F)C(F)(F)F
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
38701-74-5
C3H2F6O
170.05
MFCD00037559
BYEAHWXPCBROCE-AWPANEGFSA-N
2724852
2-deuterio-2-deuteriooxy-1,1,1,3,3,3-hexafluoropropane
[2H]OC([2H])(C(F)(F)F)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 1.6400g/mL
Deuterium (D) 99.0% min.
Quantity 1 g
Linear Formula CF3CD(OD)CF3
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol-d2, 1,1,1,3,3,3-hexafluoro 2-2h propan-2-2h ol, hexafluoro 2 h propan-2-2 h ol, hexafluoro 2h propan-2-2h ol, 1,1,1,3,3,3-hexafluoro-2-propanol-d2 hfip-d2, 1,1,1,3,3,3-hexafluoro-2-propanol-d2, 99 atom % d
Formula Weight 170.05
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropanol-d2, 99+ atom% D
compliance-icons
Product Identifier
  • 1,1,1,3,3,3-Hexafluoroisopropanol-d2
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'