missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol-d{2}, 98%(Isotopic), Thermo Scientific Chemicals

Catalog Number 11326079
Click to view available options
Quantity:
1 g
5 x 1 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
38701-74-5
C3H2F6O
170.05
MFCD00037559
BYEAHWXPCBROCE-AWPANEGFSA-N
2724852
[2H]OC([2H])(C(F)(F)F)C(F)(F)F
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
38701-74-5
C3H2F6O
170.05
MFCD00037559
BYEAHWXPCBROCE-AWPANEGFSA-N
2724852
[2H]OC([2H])(C(F)(F)F)C(F)(F)F

1,1,1,3,3,3-Hexafluoro-2-propanol-d{2} is a stable isotope used for biochemical research. It is also used as an intermediate in chemical research and in pharmaceutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 1.60 g/mL
Packaging Prescored Ampule
Sensitivity Moisture
Quantity 5 x 1 g
Melting Point −4°C
Boiling Point 59°C
UN Number UN1760
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol-d2, 1,1,1,3,3,3-hexafluoro 2-2h propan-2-2h ol, hexafluoro 2 h propan-2-2 h ol, hexafluoro 2h propan-2-2h ol, 1,1,1,3,3,3-hexafluoro-2-propanol-d2 hfip-d2, 1,1,1,3,3,3-hexafluoro-2-propanol-d2, 99 atom % d
Solubility Information Fully miscible in water.
IUPAC Name hexafluoro(²H)propan-2-(²H)ol
Formula Weight 170.05
Percent Purity 98%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol-d2, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'