missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, 99+%, Thermo Scientific Chemicals

Catalog Number p-7020700
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
920-66-1
C3H2F6O
168.038
MFCD00011651
BYEAHWXPCBROCE-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
13529
CHEBI:63104
1,1,1,3,3,3-hexafluoropropan-2-ol
C(C(F)(F)F)(C(F)(F)F)O
This item is not returnable. View return policy
920-66-1
C3H2F6O
168.038
MFCD00011651
BYEAHWXPCBROCE-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
13529
CHEBI:63104
1,1,1,3,3,3-hexafluoropropan-2-ol
C(C(F)(F)F)(C(F)(F)F)O

1,1,1,3,3,3-Hexafluoro-2-propanol is used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials. It plays an important role in the Friedel-Crafts reactions due to its polarity and high ionizing power. It finds use in sample preparation for GCMS. It also catalyzes the epoxidation of cyclooctene and 1-octene with hydrogen peroxide. It is used as a solvent for many reactions and to dissolve peptides. It is also employed to produce high-end chemicals like fluorinated surfactants, fluorinated emulsifier and fluorinated medicine. It acts as a cleaner in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,3,3,3-Hexafluoro-2-propanol is used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials. It plays an important role in the Friedel-Crafts reactions due to its polarity and high ionizing power. It finds use in sample preparation for GCMS. It also catalyzes the epoxidation of cyclooctene and 1-octene with hydrogen peroxide. It is used as a solvent for many reactions and to dissolve peptides. It is also employed to produce high-end chemicals like fluorinated surfactants, fluorinated emulsifier and fluorinated medicine. It acts as a cleaner in electronic industry.

Solubility
Miscible with water, chloroform, ether and acetone.

Notes
Incompatible with strong oxidizing agents, strong acids, strong bases, aluminum, zinc and alkali metals.
TRUSTED_SUSTAINABILITY
Melting Point -4°C
Density 1.619
Boiling Point 59°C to 60°C
Linear Formula CF3CH(OH)CF3
Refractive Index 1.275
Quantity 5 g
UN Number UN1760
Beilstein 1841007
Solubility Information Miscible with water,chloroform,ether and acetone.
Formula Weight 168.04
Percent Purity ≥99%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,1,3,3,3,-Hexafluoro-2-propanol
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 4
Hazard Statement
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'