missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals

Catalog Number p-3211286
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
920-66-1
MFCD00011651
BYEAHWXPCBROCE-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
13529
CHEBI:63104
1,1,1,3,3,3-hexafluoropropan-2-ol
C(C(F)(F)F)(C(F)(F)F)O
920-66-1
MFCD00011651
BYEAHWXPCBROCE-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
13529
CHEBI:63104
1,1,1,3,3,3-hexafluoropropan-2-ol
C(C(F)(F)F)(C(F)(F)F)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.009 max. at 240nm, 0.046 max. at 200nm, 0.097 max. at 195nm, 0.301 max. at 190nm
CAS Min % 99.85
CAS Max % 100.0
Melting Point -4.0°C
Density 1.5960g/mL
Boiling Point 59.0°C
Flash Point >100°C
Assay Percent Range 99.85% min. (GC)
Packaging Glass bottle
Molecular Formula C3H2F6O
Linear Formula (CF3)2CHOH
Refractive Index 1.2740 to 1.2770
Quantity 10 g
Residue after Evaporation 0.001% max.
Specific Gravity 1.596
Solubility Information Solubility in water: 1000g/L (25°C)
Molecular Weight (g/mol) 168.04
Viscosity 1.65 mPa.s (20°C)
Formula Weight 168.04
Percent Purity 99.9%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1,1,3,3,3-Hexafluoro-2-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'