missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals
68.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
Specifications
Absorbance | (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.009 max. at 240nm, 0.046 max. at 200nm, 0.097 max. at 195nm, 0.301 max. at 190nm |
CAS Min % | 99.85 |
CAS Max % | 100.0 |
Melting Point | -4.0°C |
Density | 1.5960g/mL |
Boiling Point | 59.0°C |
Flash Point | >100°C |
Assay Percent Range | 99.85% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C3H2F6O |
Show More |
Safety and Handling
Product Identifier
- 1,1,1,3,3,3-Hexafluoro-2-propanol
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 C
- Specific target organ toxicity after repeated exposure Category 2
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H361d-Suspected of damaging the unborn child.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- H373-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'