missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,2,2-Pentafluoro-3-iodopropane, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
1,1,1,2,2-Pentafluoro-3-iodopropane is used to produce 1,1,1,2,2-pentafluoro-propane by heating. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1,1,2,2-Pentafluoro-3-iodopropane is used to produce 1,1,1,2,2-pentafluoro-propane by heating. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with strong oxidizing agents.
1,1,1,2,2-Pentafluoro-3-iodopropane is used to produce 1,1,1,2,2-pentafluoro-propane by heating. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with strong oxidizing agents.
Specifications
Boiling Point | 70°C to 71°C |
Refractive Index | 1.373 |
Quantity | 1 g |
Beilstein | 1700278 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 259.95 |
Percent Purity | 96% |
Chemical Name or Material | 1,1,1,2,2-Pentafluoro-3-iodopropane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'