missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 97%, Thermo Scientific™

Catalog Number p-3758864
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2043-57-4
C8H4F13I
474.004
MFCD00039410
NVVZEKTVIXIUKW-UHFFFAOYSA-N
1h,1h,2h,2h-perfluorooctyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodo, 1-iodo-1h,1h,2h,2h-perfluorooctane, 1,1,2,2-tetrahydroperfluorooctyliodide, 1,1,2,2-tetrahydroperfluorooctyl iodide, 2-perfluorohexyl ethyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-iodooctane, 1h,1h,2h,2h-tridecafluoro-n-octyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl iodide, acmc-1ccsn
74888
1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodooctane
C(CI)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
This item is not returnable. View return policy
2043-57-4
C8H4F13I
474.004
MFCD00039410
NVVZEKTVIXIUKW-UHFFFAOYSA-N
1h,1h,2h,2h-perfluorooctyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodo, 1-iodo-1h,1h,2h,2h-perfluorooctane, 1,1,2,2-tetrahydroperfluorooctyliodide, 1,1,2,2-tetrahydroperfluorooctyl iodide, 2-perfluorohexyl ethyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-iodooctane, 1h,1h,2h,2h-tridecafluoro-n-octyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl iodide, acmc-1ccsn
74888
1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodooctane
C(CI)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Density 1.9340g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ICH2CH2(CF2)5CF3
Refractive Index 1.3590 to 1.3610 (20°C, 589nm)
Quantity 5 g
Formula Weight 474
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'