missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 96 Well 0.8mL Polypropylene Deepwell Storage Plate

0.8 mL deepwell storage plates for cell culture or sample storage applications.

Brand:  Thermo Scientific™ AB-0765/K

Code : 03

Additional Details : Weight : 3.90000kg

 View more versions of this product

Product Code. 10029522

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

ABgene storage and assay plates are manufactured and in a Class 100,000 cleanroom environment to ensure that each plate is contamination-free. Plates are made using virgin medical grade polypropylene and have highly polished surfaces making them low binding and chemically resistant. Rigid side walls and industry standard footprint make plates suitable for automation and robotic handling, as well as for low-throughput applications.
  • 96 Round wells with 800μL maximum well volume
  • Max working volume: Film sealing = 0.8mL, Individual Cap Sealing = 0.7mL, Mat Cap Sealing = 0.65mL
  • Raised rim design around each well for optimal sealing and reduced risk of cross-contamination
  • Suitable for use with DMSO, Ethanol, Isopropanol
  • Alphanumeric grid-referencing
  • Designed according to SBS footprint standards for robotic compatibility
  • Wide working temperature range from -196° to +121°C (-321° to +250°F)
  • Can be bar coded for improved sample tracking, please inquire
  • Autoclavable (15 min. at 121°C), safe for gamma irradiation sterilization
  • Max RCF: 200xG (0.65mL/well)
  • Manufactured from virgin medical grade polypropylene and certified RNase, DNase and endotoxin free

Compatible with:

Sealing options include: Adhesive Sealing (0.8mL working volume); Heat sealing (0.8mL working volume); Storage Plate Cap Strips (0.75mL working volume); 96 Cap Sealing Mats (0.65mL working volume)

Specifications

Specifications

Certified free from human DNA, RNases, and DNases.
Black
Conical
Black
800┬ÁL
50
Videos
Documentation

Technical Documents

Alternative Products