Learn More
Fisher Chemical 0.5LT ETHANOL 95% V/V DP A.R., MA: E/0550/08charged at the current rate for analysis CertiFied
Brand: Fisher Chemical E/0550/08
0.5LT ETHANOL 95% V/V DP A.R., MA: E/0550/08charged at the current rate for analysis CertiFied
Description
Safety and Handling
- Ethanol (95% v/v)
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.