accessibility menu, dialog, popup

UserName

CAS RN 99848-43-8

H3COBrONH2

CAS RN 99848-43-8

IUPAC Name: 2-bromo-5-methoxybenzamide
Synonyms: 2-bromo-5-methoxybenzamide
Molecular Weight (g/mol): 230.06
Molecular Formula: C8H8BrNO2
InChi Key: CMTKLJYDFPAPJS-UHFFFAOYSA-N
SMILES: COC1=CC(C(N)=O)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results