accessibility menu, dialog, popup

UserName

CAS RN 3232-84-6

ONHNHOHN

CAS RN 3232-84-6

IUPAC Name: 1,2,4-triazolidine-3,5-dione
Synonyms: 1,2,4-triazolidine-3,5-dione
Molecular Weight (g/mol): 101.07
Molecular Formula: C2H3N3O2
InChi Key: UDATXMIGEVPXTR-UHFFFAOYSA-N
SMILES: O=C1NNC(=O)N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results