accessibility menu, dialog, popup

UserName

CAS RN 126163-30-2

FFHO

CAS RN 126163-30-2

IUPAC Name: 2-(2,3-difluorophenyl)ethan-1-ol
Synonyms: 2-(2,3-difluorophenyl)ethanol
Molecular Weight (g/mol): 158.15
Molecular Formula: C8H8F2O
InChi Key: RBNNHALDGIKSBZ-UHFFFAOYSA-N
SMILES: OCCC1=C(F)C(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results