accessibility menu, dialog, popup

UserName

CAS RN 125116-23-6

CH3H3CClHONNN

CAS RN 125116-23-6

IUPAC Name: 5-[(4-chlorophenyl)methyl]-2,2-dimethyl-1-[(1H-1,2,4-triazol-1-yl)methyl]cyclopentan-1-ol
Synonyms: metconazole metconazole (iso)
Molecular Weight (g/mol): 319.83
Molecular Formula: C17H22ClN3O
InChi Key: XWPZUHJBOLQNMN-UHFFFAOYNA-N
SMILES: CC1(C)CCC(CC2=CC=C(Cl)C=C2)C1(O)CN1C=NC=N1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (4)
Concentration 
  • (2)
  • (2)
Solvent or Matrix 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)